Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zmynd12
Entrez Gene ID
492338
Gene Name
zinc finger, MYND-type containing 12
Gene Aliases
CH73-261F6.1, zgc:92280, zymnd12
Chromosome Location
Chr.8: 516469 - 531286 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007304.1 NP_001007305.1 2 - 3 382 72
XM_021478243.1 XP_021333918.1 2 - 3 453 72
GenBank mRNA BC085655.1 - 2 - 3 382 72

View Full Product Details