Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fli1rs
Entrez Gene ID
386723
Gene Name
Fli-1 proto-oncogene, ETS transcription factor-related sequence
Gene Aliases
fli1b, zgc:5777174
Chromosome Location
Chr.16: 42017966 - 42046881 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008780.1 NP_001008780.1 7 - 8 908 92
XM_005158200.3 XP_005158257.1 7 - 8 984 92
XM_005158202.4 XP_005158259.1 7 - 8 900 92
XM_005158203.4 XP_005158260.1 7 - 8 897 92
XM_005158205.4 XP_005158262.1 7 - 8 897 92
XM_009292528.3 XP_009290803.1 7 - 8 1016 92
XM_009292529.3 XP_009290804.1 7 - 8 1013 92
XM_009292530.3 XP_009290805.1 7 - 8 1015 92
XM_009292531.3 XP_009290806.1 7 - 8 1012 92
GenBank mRNA AY839950.1 - 7 - 8 908 92
BC055627.1 - 7 - 8 829 92

View Full Product Details