Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dcdc2b
Entrez Gene ID
563259
Gene Name
doublecortin domain containing 2B
Gene Aliases
zgc:123267
Chromosome Location
Chr.13: 33301899 - 33314758 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037689.2 NP_001032778.2 7 - 8 811 107
XM_017358759.2 XP_017214248.1 9 - 10 1177 107
XM_017358760.2 XP_017214249.1 9 - 10 1151 107
XM_017358761.2 XP_017214250.1 9 - 10 1164 107
XM_017358762.2 XP_017214251.1 9 - 10 1138 107
XM_017358763.2 XP_017214252.1 9 - 10 1177 107
XM_017358764.2 XP_017214253.1 9 - 10 1138 107
XM_017358765.2 XP_017214254.1 9 - 10 1177 107
XM_021480609.1 XP_021336284.1 8 - 9 2551 107
GenBank mRNA BC108062.1 - 7 - 8 822 107

View Full Product Details