Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btr32
Entrez Gene ID
678629
Gene Name
bloodthirsty-related gene family, member 32
Gene Aliases
zgc:136954
Chromosome Location
Chr.23: 46163188 - 46180011 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021470074.1 XP_021325749.1 1 - 2 608 84
GenBank mRNA BC115345.1 - 2 - 3 597 84

View Full Product Details