Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scn1lab
Entrez Gene ID
559447
Gene Name
sodium channel, voltage-gated, type I like, alpha b
Gene Aliases
-
Chromosome Location
Chr.6: 10246549 - 10321670 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044895.2 NP_001038360.2 10 - 11 1681 87
XM_005165770.4 XP_005165827.1 11 - 12 2302 87
XM_009302186.3 XP_009300461.1 11 - 12 1949 87
XM_021476883.1 XP_021332558.1 10 - 11 1628 87
XM_021476884.1 XP_021332559.1 10 - 11 1656 87
XM_021476885.1 XP_021332560.1 10 - 11 1628 87
XM_021476886.1 XP_021332561.1 10 - 11 1628 87
XM_021476887.1 XP_021332562.1 10 - 11 1628 87
GenBank mRNA DQ149504.1 - 10 - 11 1628 87

View Full Product Details