Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eif3eb
Entrez Gene ID
561049
Gene Name
eukaryotic translation initiation factor 3, subunit E, b
Gene Aliases
eif3s6b, im:7147439
Chromosome Location
Chr.19: 22624269 - 22664172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001362621.1 NP_001349550.1 10 - 11 1093 125

View Full Product Details