Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc45a2
Entrez Gene ID
558311
Gene Name
solute carrier family 45 member 2
Gene Aliases
aim1, im:7138762
Chromosome Location
Chr.21: 19445942 - 19484289 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110377.1 NP_001103847.1 4 - 5 1157 69
GenBank mRNA BC154627.1 - 4 - 5 1157 69

View Full Product Details