Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clic3
Entrez Gene ID
84040
Gene Name
chloride intracellular channel 3
Gene Aliases
clica, zgc:77835
Chromosome Location
Chr.21: 13663656 - 13672190 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199524.1 NP_955818.1 4 - 5 605 131
XM_005161114.4 XP_005161171.1 5 - 6 930 131
GenBank mRNA BC062860.1 - 4 - 5 622 131

View Full Product Details