Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lsm4
Entrez Gene ID
393669
Gene Name
LSM4 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
zgc:73057, zgc:77848
Chromosome Location
Chr.11: 6859876 - 6868240 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200696.1 NP_956990.1 5 - 6 548 94
GenBank mRNA BC059435.1 - 5 - 6 548 94
BC062285.1 - 5 - 6 529 94

View Full Product Details