Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
veph1
Entrez Gene ID
405756
Gene Name
ventricular zone expressed PH domain-containing 1
Gene Aliases
veph, zVeph-A
Chromosome Location
Chr.18: 115970 - 274820 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212831.1 NP_997996.1 4 - 5 731 65
GenBank mRNA AB085188.1 - 4 - 5 731 65
BC134890.1 - 4 - 5 711 65

View Full Product Details