Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cryabb
Entrez Gene ID
436943
Gene Name
crystallin, alpha B, b
Gene Aliases
cryab2, fe37f08, wu:fe37f08, zgc:91937
Chromosome Location
Chr.5: 258933 - 263246 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002670.2 NP_001002670.1 2 - 3 445 123
XM_021476081.1 XP_021331756.1 2 - 3 904 123
GenBank mRNA AY939876.1 - 2 - 3 314 123
BC076518.1 - 2 - 3 433 123
BC163968.1 - 2 - 3 323 123
DQ113417.1 - 2 - 3 445 123

View Full Product Details