Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
optc
Entrez Gene ID
445189
Gene Name
opticin
Gene Aliases
dspg3, optcl, wu:fb81a05, zgc:101047
Chromosome Location
Chr.11: 23205828 - 23219361 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003583.2 NP_001003583.2 6 - 7 856 143
XM_005172873.4 XP_005172930.1 6 - 7 890 143
GenBank mRNA BC078270.1 - 6 - 7 892 143
CU468787.1 - 6 - 7 797 143
DQ224359.1 - 6 - 7 859 143

View Full Product Details