Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zmat5
Entrez Gene ID
445314
Gene Name
zinc finger, matrin-type 5
Gene Aliases
zgc:101022
Chromosome Location
Chr.5: 32076109 - 32080192 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003771.1 NP_001003771.1 3 - 4 392 83
GenBank mRNA BC079486.1 - 3 - 4 392 83

View Full Product Details