Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hif1al2
Entrez Gene ID
497283
Gene Name
hypoxia inducible factor 1 subunit alpha, like 2
Gene Aliases
hif1al, zgc:112119
Chromosome Location
Chr.21: 27327424 - 27339396 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012371.2 NP_001012371.2 4 - 5 608 157
XM_005161350.4 XP_005161407.1 5 - 6 945 157
XM_021468817.1 XP_021324492.1 6 - 7 821 157
GenBank mRNA AY835381.1 - 4 - 5 521 157
BC093209.1 - 4 - 5 609 157

View Full Product Details