Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bcl2a
Entrez Gene ID
570772
Gene Name
BCL2 apoptosis regulator a
Gene Aliases
bcl2
Chromosome Location
Chr.24: 28137210 - 28228917 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030253.2 NP_001025424.1 1 - 2 610 67
GenBank mRNA AY695820.1 - 1 - 2 555 67
BC133848.1 - 1 - 2 610 67

View Full Product Details