Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ahrrb
Entrez Gene ID
565319
Gene Name
aryl-hydrocarbon receptor repressor b
Gene Aliases
ahrr2
Chromosome Location
Chr.2: 247514 - 297658 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033920.1 NP_001029092.1 5 - 6 570 84
GenBank mRNA AY928204.1 - 5 - 6 570 84
BC163123.1 - 5 - 6 570 84

View Full Product Details