Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppm1h
Entrez Gene ID
768291
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1H
Gene Aliases
zgc:153678
Chromosome Location
Chr.25: 1592094 - 1624974 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077455.1 NP_001070923.1 2 - 3 394 65
GenBank mRNA BC124421.1 - 2 - 3 394 65

View Full Product Details