Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcmt
Entrez Gene ID
30751
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase
Gene Aliases
etID309741.20, fj13d06, pimt, wu:fj13d06
Chromosome Location
Chr.20: 1293985 - 1314555 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131465.3 NP_571540.1 7 - 8 757 78
GenBank mRNA BC049067.1 - 7 - 8 753 78
BC075735.1 - 7 - 8 725 78
U37434.1 - 7 - 8 716 78

View Full Product Details