Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rpe65a
Entrez Gene ID
393724
Gene Name
retinoid isomerohydrolase RPE65 a
Gene Aliases
RPE56a, rpepa, zgc:73213
Chromosome Location
Chr.2: 1428617 - 1443411 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200751.1 NP_957045.1 11 - 12 1272 93
GenBank mRNA AY646886.1 - 10 - 11 1239 93
BC059559.1 - 11 - 12 1272 93

View Full Product Details