Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nucb2a
Entrez Gene ID
373078
Gene Name
nucleobindin 2a
Gene Aliases
cb638, nucb2, wu:fa03d10
Chromosome Location
Chr.7: 56540 - 69730 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201493.1 NP_958901.1 5 - 6 594 68
GenBank mRNA BC046077.1 - 5 - 6 594 68
BC065437.1 - 5 - 6 560 68

View Full Product Details