Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbl2
Entrez Gene ID
337068
Gene Name
transducin beta like 2
Gene Aliases
wu:fa96f08
Chromosome Location
Chr.10: 33254710 - 33265677 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212767.1 NP_997932.1 5 - 6 1017 112
XM_005172641.4 XP_005172698.1 6 - 7 112
GenBank mRNA BC065682.1 - 5 - 6 1017 112
BC153607.1 - 5 - 6 112

View Full Product Details