Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cab39
Entrez Gene ID
415235
Gene Name
calcium binding protein 39
Gene Aliases
wu:fc06a03, zgc:86716
Chromosome Location
Chr.15: 39977461 - 40002930 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002145.1 NP_001002145.1 7 - 8 897 119
GenBank mRNA BC071393.1 - 7 - 8 897 119

View Full Product Details