Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nop16
Entrez Gene ID
436807
Gene Name
NOP16 nucleolar protein homolog (yeast)
Gene Aliases
zgc:92910
Chromosome Location
Chr.21: 36612439 - 36619599 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002534.1 NP_001002534.1 3 - 4 329 127
GenBank mRNA BC076362.1 - 3 - 4 329 127

View Full Product Details