Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfr1a
Entrez Gene ID
494477
Gene Name
transferrin receptor 1a
Gene Aliases
-
Chromosome Location
Chr.2: 4844080 - 4874960 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009917.1 NP_001009917.1 1 - 2 36 61
XM_005163666.4 XP_005163723.1 2 - 3 137 61
GenBank mRNA AY649363.1 - 1 - 2 36 61
BC162721.1 - 1 - 2 63 61

View Full Product Details