Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
elp4
Entrez Gene ID
550331
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
fi38c09, wu:fi38c09, zgc:112389
Chromosome Location
Chr.7: 15906222 - 16033854 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017638.1 NP_001017638.1 7 - 8 898 91
XM_009302967.3 XP_009301242.1 7 - 8 905 91
GenBank mRNA BC093280.1 - 7 - 8 898 91

View Full Product Details