Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vbp1
Entrez Gene ID
553651
Gene Name
von Hippel-Lindau binding protein 1
Gene Aliases
im:7140038, zgc:110686
Chromosome Location
Chr.7: 23887728 - 23894685 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020624.1 NP_001018460.1 4 - 5 393 78
GenBank mRNA BC095370.1 - 4 - 5 393 78

View Full Product Details