Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pglyrp2
Entrez Gene ID
568634
Gene Name
peptidoglycan recognition protein 2
Gene Aliases
pgrp-l
Chromosome Location
Chr.6: 8490657 - 8499017 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045166.1 NP_001038631.1 4 - 5 1346 88
XM_021476909.1 XP_021332584.1 4 - 5 1624 88
GenBank mRNA DQ447202.1 - 4 - 5 1346 88
DQ482738.1 - 4 - 5 1283 88

View Full Product Details