Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
m17
Entrez Gene ID
555717
Gene Name
IL-6 subfamily cytokine M17
Gene Aliases
lif, si:dkeyp-15f12.5
Chromosome Location
Chr.5: 32155227 - 32158198 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079833.1 NP_001073302.1 1 - 2 41 76
GenBank mRNA AB271233.1 - 1 - 2 41 76

View Full Product Details