Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekhm1
Entrez Gene ID
564048
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 1
Gene Aliases
wu:fc17c08, wu:fc23c01, wu:fc29h10, zgc:158281
Chromosome Location
Chr.12: 102409 - 144473 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001327851.1 NP_001314780.1 10 - 11 3054 97
NM_001327852.1 NP_001314781.1 9 - 10 2415 97
XM_021480128.1 XP_021335803.1 10 - 11 3201 97
GenBank mRNA BC133733.1 - 9 - 10 2366 97

View Full Product Details