Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thap7
Entrez Gene ID
100006965
Gene Name
THAP domain containing 7
Gene Aliases
im:7136959, si:ch211-282e16.1
Chromosome Location
Chr.19: 24129414 - 24136233 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103198.1 NP_001096668.1 5 - 6 519 83
XM_021467964.1 XP_021323639.1 3 - 4 298 83
GenBank mRNA BC171394.1 - 4 - 5 388 83

View Full Product Details