Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
h1m
Entrez Gene ID
327403
Gene Name
linker histone H1M
Gene Aliases
cb812, fi06c02, wu:fd14d08, wu:fi06c02
Chromosome Location
Chr.5: 68327148 - 68332660 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183071.2 NP_898894.2 1 - 2 145 95
GenBank mRNA BC083463.1 - 1 - 2 127 95
BC097052.1 - 1 - 2 145 95

View Full Product Details