Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mpp5a
Entrez Gene ID
252845
Gene Name
membrane protein, palmitoylated 5a (MAGUK p55 subfamily member 5)
Gene Aliases
mpp5, nok
Chromosome Location
Chr.17: 34186605 - 34239519 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194363.1 NP_919344.1 5 - 6 1077 108
XM_009293174.3 XP_009291449.1 6 - 7 1179 108
GenBank mRNA AF510111.1 - 5 - 6 1077 108
BC163251.1 - 3 - 4 811 108
BC163277.1 - 3 - 4 811 108

View Full Product Details