Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
serpinb1l3
Entrez Gene ID
335119
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 1, like 3
Gene Aliases
fk71e06, wu:fk71e06, zgc:64178
Chromosome Location
Chr.24: 42148140 - 42157919 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199936.1 NP_956230.1 6 - 7 825 112
XM_005174205.4 XP_005174262.1 6 - 7 1017 112
GenBank mRNA BC053300.1 - 6 - 7 825 112

View Full Product Details