Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ivns1abpb
Entrez Gene ID
334334
Gene Name
influenza virus NS1A binding protein b
Gene Aliases
fi13f08, fi41c09, wu:fi13f08, wu:fi41c09
Chromosome Location
Chr.2: 23379401 - 23390742 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201483.1 NP_958891.1 11 - 12 1523 62
XM_021479057.1 XP_021334732.1 5 - 6 981 62
GenBank mRNA BC045449.1 - 11 - 12 1523 62

View Full Product Details