Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pard6gb
Entrez Gene ID
80959
Gene Name
par-6 family cell polarity regulator gamma b
Gene Aliases
PARD6G, cb182, id:ibd2873, sb:cb182, wu:fc33c12
Chromosome Location
Chr.19: 25699329 - 25772843 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212563.1 NP_997728.1 2 - 3 307 61
GenBank mRNA AY391446.1 - 2 - 3 307 61
BC059186.1 - 2 - 3 444 61

View Full Product Details