The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003783.2 | NP_001003783.1 | 6 - 7 | 797 | 134 |
GenBank mRNA | BC077140.1 | - | 6 - 7 | 801 | 134 |