Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxl22
Entrez Gene ID
503591
Gene Name
F-box and leucine-rich repeat protein 22
Gene Aliases
zgc:110743
Chromosome Location
Chr.25: 26727109 - 26735957 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013279.2 NP_001013297.1 1 - 2 408 62
GenBank mRNA BC090262.1 - 1 - 2 412 62

View Full Product Details