Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm8a
Entrez Gene ID
503767
Gene Name
RNA binding motif protein 8A
Gene Aliases
fb34f03, wu:fb34f03, zgc:110525
Chromosome Location
Chr.16: 45922224 - 45930077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013345.1 NP_001013363.1 4 - 5 374 90
GenBank mRNA BC090261.1 - 4 - 5 374 90

View Full Product Details