Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2y3
Entrez Gene ID
368352
Gene Name
cytochrome P450, family 2, subfamily Y, polypeptide 3
Gene Aliases
im:6903403, sb:cb637, zgc:110692
Chromosome Location
Chr.15: 26021 - 32151 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020822.1 NP_001018658.1 5 - 6 839 139
GenBank mRNA AY927850.1 - 5 - 6 835 139
BC095374.1 - 5 - 6 839 139

View Full Product Details