Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfap2b
Entrez Gene ID
550545
Gene Name
transcription factor AP-2 beta
Gene Aliases
si:dkey-5e16.1, zgc:110163
Chromosome Location
Chr.20: 47688719 - 47706993 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024665.1 NP_001019836.1 4 - 5 991 66
XM_005160883.4 XP_005160940.1 4 - 5 959 66
GenBank mRNA DQ060246.1 - 4 - 5 991 66

View Full Product Details