Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vtg2
Entrez Gene ID
559931
Gene Name
vitellogenin 2
Gene Aliases
si:dkey-4c23.2, vg2, zfVtg2
Chromosome Location
Chr.22: 24869690 - 24880824 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044913.1 NP_001038378.1 16 - 17 2270 94
GenBank mRNA BC133137.1 - 16 - 17 2344 94

View Full Product Details