Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rad21l1
Entrez Gene ID
571082
Gene Name
RAD21 cohesin complex component like 1
Gene Aliases
CH1073-428E6.1, zgc:154013
Chromosome Location
Chr.8: 49117518 - 49140645 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080050.1 NP_001073519.1 4 - 5 437 87
XM_021478297.1 XP_021333972.1 3 - 4 468 87
GenBank mRNA BC125933.1 - 4 - 5 437 87

View Full Product Details