Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnmt3bb.2
Entrez Gene ID
30659
Gene Name
DNA (cytosine-5-)-methyltransferase 3 beta, duplicate b.2
Gene Aliases
Zmt3, dnmt3, wu:fa11d06, wu:fb73b09
Chromosome Location
Chr.23: 16983363 - 17004783 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131386.1 NP_571461.1 13 - 14 3219 100
GenBank mRNA AB196914.1 - 13 - 14 3219 100
AF135438.1 - 9 - 10 1202 100
BC162467.1 - 13 - 14 3108 100

View Full Product Details