Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ebf2
Entrez Gene ID
30692
Gene Name
EBF transcription factor 2
Gene Aliases
Zcoe2, coe2
Chromosome Location
Chr.5: 67390554 - 67439831 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131418.2 NP_571493.2 4 - 5 858 72
XM_005165618.4 XP_005165675.1 4 - 5 910 72
GenBank mRNA AF072657.1 - 4 - 5 818 72
BC162504.1 - 4 - 5 667 72

View Full Product Details