Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ucp1
Entrez Gene ID
83908
Gene Name
uncoupling protein 1
Gene Aliases
UCP3, UCPx, fb62c08, ucp4, wu:fb62c08, zgc:55824, zgc:77603
Chromosome Location
Chr.1: 53374488 - 53388911 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199523.2 NP_955817.1 2 - 3 200 66
GenBank mRNA AY398428.1 - 2 - 3 208 66
BC045464.1 - 2 - 3 208 66
BC063945.1 - 2 - 3 200 66
BC075906.1 - 2 - 3 194 66
BC164087.1 - 1 - 2 164 66

View Full Product Details