Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arpc1a
Entrez Gene ID
394243
Gene Name
actin related protein 2/3 complex, subunit 1A
Gene Aliases
wu:fk93e03
Chromosome Location
Chr.3: 40796111 - 40804455 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201343.2 NP_958500.1 2 - 3 262 60
GenBank mRNA AY398313.1 - 2 - 3 260 60
BC048036.1 - 2 - 3 253 60
BC164199.1 - 1 - 2 99 60

View Full Product Details