Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnf8
Entrez Gene ID
404050
Gene Name
ring finger protein 8, E3 ubiquitin protein ligase
Gene Aliases
fc66f09, wu:fc66f09, zgc:55936, zgc:77313
Chromosome Location
Chr.20: 51363686 - 51386564 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205553.1 NP_991116.1 6 - 7 1149 123
NM_205766.1 NP_991329.1 6 - 7 1202 123
GenBank mRNA BC044545.1 - 6 - 7 1202 123
BC065643.1 - 6 - 7 1149 123

View Full Product Details