Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cryba2a
Entrez Gene ID
415139
Gene Name
crystallin, beta A2a
Gene Aliases
zgc:86610
Chromosome Location
Chr.6: 13780210 - 13783634 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002049.1 NP_001002049.1 2 - 3 225 78
GenBank mRNA BC071312.1 - 2 - 3 225 78
DQ000463.1 - 2 - 3 214 78

View Full Product Details