Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab11fip4a
Entrez Gene ID
436806
Gene Name
RAB11 family interacting protein 4 (class II) a
Gene Aliases
rab11fip4, zgc:92911
Chromosome Location
Chr.3: 36018098 - 36115355 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002533.2 NP_001002533.2 11 - 12 1409 106
XM_009299531.3 XP_009297806.1 11 - 12 1407 106
XM_009299532.3 XP_009297807.1 9 - 10 1317 106
GenBank mRNA AB208638.1 - 11 - 12 1409 106
AB208639.1 - 9 - 10 1281 106

View Full Product Details