Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc1a1
Entrez Gene ID
436939
Gene Name
solute carrier family 1 member 1
Gene Aliases
EAAT3, SLC1A2a, zgc:91959
Chromosome Location
Chr.7: 69496293 - 69521477 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002666.1 NP_001002666.2 10 - 11 1322 59
GenBank mRNA BC076553.1 - 10 - 11 1322 59
BC163973.1 - 10 - 11 1236 59
HM138690.1 - 10 - 11 1208 59

View Full Product Details